Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,4-di-tert-butylphenyl) hydrogen phosphate 97%

Supplier: AMBEED, INC
Warning

A1150398-100MG A1150398-1G A1150398-250MG A1150398-5G
76674-268EA 29 USD InStock
76674-268 76674-270 76674-272 76674-274
Bis(2,4-di-tert-butylphenyl) hydrogen phosphate 97%
Bis(2,4-di-tert-butylphenyl) hydrogen phosphate
Precaución: for Research Use Only


Formula: C₂₈H₄₃O₄P
MDL Number: MFCD29059882
CAS Number: 69284-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty