Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis (2,2'-bipyridyl) (2,2'-bipyrimidine) ruthenium hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A1150986-1G A1150986-250MG A1150986-100MG
76674-348EA 556 USD InStock
76674-348 76674-350 77620-124
Bis (2,2'-bipyridyl) (2,2'-bipyrimidine) ruthenium hexafluorophosphate 98%
Bis (2,2'-bipyridyl) (2,2'-bipyrimidine) ruthenium hexafluorophosphate
Precaución: for Research Use Only


Formula: C₂₈H₂₂F₁₂N₈P₂Ru
MDL Number: MFCD32856149
CAS Number: 65013-23-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty