Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fumaraldehydebis(dimethyl acetal) 90+%

Supplier: AMBEED, INC
Warning

A1151619-1G A1151619-250MG A1151619-25G A1151619-5G
76674-378EA 36 USD InStock
76674-378 76674-380 76674-382 76674-384
Fumaraldehydebis(dimethyl acetal) 90+%
Fumaraldehydebis(dimethyl acetal)
Precaución: for Research Use Only


Formula: C₈H₁₆O₄
MDL Number: MFCD00075507
CAS Number: 6068-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty