Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(6-Morpholino-1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)hexanoic acid 95%

Supplier: AMBEED, INC
Warning

A1152101-100MG A1152101-1G A1152101-250MG
76674-424EA 59 USD InStock
76674-424 76674-426 76674-428
6-(6-Morpholino-1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)hexanoic acid 95%
6-(6-Morpholino-1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)hexanoic acid
Precaución: for Research Use Only


Formula: C₂₂H₂₄N₂O₅
MDL Number: MFCD02916602
CAS Number: 423146-25-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty