Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dibenzylamino)-2,2-difluoropropanoic acid 95%

Supplier: AMBEED, INC
Warning

A1152146-100MG A1152146-1G A1152146-250MG
76674-432EA 7 USD InStock
76674-432 76674-434 76674-436
3-(Dibenzylamino)-2,2-difluoropropanoic acid 95%
3-(Dibenzylamino)-2,2-difluoropropanoic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₇F₂NO₂
MDL Number: MFCD32638827
CAS Number: 890146-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty