Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3R,6R)-1-tert-Butyl 3-methyl 6-methylpiperazine-1,3-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A1152532-1G A1152532-250MG
76674-462EA 57 USD InStock
76674-462 77620-144
(3R,6R)-1-tert-Butyl 3-methyl 6-methylpiperazine-1,3-dicarboxylate 97%
(3R,6R)-1-tert-Butyl 3-methyl 6-methylpiperazine-1,3-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₂H₂₂N₂O₄
MDL Number: MFCD32644572
CAS Number: 2349395-78-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty