Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Glumetinib 99%

Supplier: AMBEED, INC
Warning

A1154012-10MG A1154012-1MG A1154012-5MG A1154012-50MG
76674-538EA 48 USD InStock
76674-538 76674-540 76674-542 77620-166
Glumetinib 99%
Glumetinib
Precaución: for Research Use Only


Formula: C₂₁H₁₇N₉O₂S
MW: 459.49 g/mol
MDL Number: MFCD31693844
CAS Number: 1642581-63-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty