Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-4-(4-Chlorophenyl)-4-oxobut-2-enoic acid 98%

Supplier: AMBEED, INC
Warning

A1154546-5G A1154546-25G
76674-562EA 35 USD InStock
76674-562 77620-172
(E)-4-(4-Chlorophenyl)-4-oxobut-2-enoic acid 98%
(E)-4-(4-Chlorophenyl)-4-oxobut-2-enoic acid
Precaución: for Research Use Only


Formula: C₁₀H₇ClO₃
MW: 210.6138 g/mol
MDL Number: MFCD00041465
CAS Number: 29582-39-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty