Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Tri-1H-pyrazol-1-yl-1,3,5-triazine 98%

Supplier: AMBEED, INC
Warning

A1154744-100MG A1154744-1G A1154744-250MG
76674-628EA 36 USD InStock
76674-628 76674-630 77620-180
2,4,6-Tri-1H-pyrazol-1-yl-1,3,5-triazine 98%
2,4,6-Tri-1H-pyrazol-1-yl-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₁₂H₉N₉
MDL Number: MFCD28336591
CAS Number: 27257-90-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty