Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Di-4-pyridinyl-4-pyridinamine 98%

Supplier: AMBEED, INC
Warning

A1154774-100MG A1154774-1G A1154774-250MG
76674-676EA 25 USD InStock
76674-676 76674-678 76674-680
N,N-Di-4-pyridinyl-4-pyridinamine 98%
N,N-Di-4-pyridinyl-4-pyridinamine
Precaución: for Research Use Only


Formula: C₁₅H₁₂N₄
CAS Number: 153467-50-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty