Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'',4''''-Nitrilotris(([1,1'-biphenyl]-3,5-dicarboxylic acid)) 98%

Supplier: AMBEED, INC
Warning

A1154790-100MG A1154790-1G A1154790-250MG
76674-730EA 20 USD InStock
76674-730 76674-732 76674-734
4,4'',4''''-Nitrilotris(([1,1'-biphenyl]-3,5-dicarboxylic acid)) 98%
4,4'',4''''-Nitrilotris(([1,1'-biphenyl]-3,5-dicarboxylic acid))
Precaución: for Research Use Only


Formula: C₄₂H₂₇NO₁₂
MW: 737.6633 g/mol
MDL Number: MFCD32641633
CAS Number: 1347748-59-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty