Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-(2H-tetrazol-5-yl)phenyl)amine 98%

Supplier: AMBEED, INC
Danger

A1154791-100MG A1154791-250MG A1154791-5G
76674-736EA 29 USD InStock
76674-736 76674-738 76674-740
Tris(4-(2H-tetrazol-5-yl)phenyl)amine 98%
Tris(4-(2H-tetrazol-5-yl)phenyl)amine
Precaución: for Research Use Only


Formula: C₂₁H₁₅N₁₃
CAS Number: 1346621-21-3
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty