Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-(5'-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 98%

Supplier: AMBEED, INC
Warning

A1154804-100MG A1154804-10G A1154804-250MG A1154804-5G
76674-772EA 13 USD InStock
76674-772 76674-774 76674-776 76674-778
2,2'-(5'-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) 98%
2,2'-(5'-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-[1,1':3',1''-terphenyl]-4,4''-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Precaución: for Research Use Only


Formula: C₄₂H₅₁B₃O₆
MDL Number: MFCD30489708
CAS Number: 1017967-97-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty