Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Propoxycinnamic acid 97%

Supplier: AMBEED, INC
Warning

A1154850-1G A1154850-25G A1154850-5G
76674-806EA 11 USD InStock
76674-806 76674-808 76674-810
4-Propoxycinnamic acid 97%
4-Propoxycinnamic acid


Formula: C₁₂H₁₄O₃
MDL Number: MFCD00995149
CAS Number: 69033-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty