Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Ethylbutyl 2-aminopropanoate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1154910-100G A1154910-1G A1154910-5G A1154910-25G
76674-818EA 421 USD InStock
76674-818 76674-820 76674-822 77620-204
(S)-2-Ethylbutyl 2-aminopropanoate hydrochloride 97%
(S)-2-Ethylbutyl 2-aminopropanoate hydrochloride
Precaución: for Research Use Only


Formula: C₉H₂₀ClNO₂
MDL Number: MFCD32660031
CAS Number: 946511-97-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty