Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(1-Phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine 98%

Supplier: AMBEED, INC
Warning

A1155016-10MG A1155016-1MG A1155016-50MG A1155016-5MG
76674-844EA 45 USD InStock
76674-844 76674-846 76674-848 76674-850
N-(1-Phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine 98%
N-(1-Phenylethyl)-2-(piperazin-1-yl)quinazolin-4-amine
Precaución: for Research Use Only


Formula: C₂₀H₂₃N₅
MDL Number: MFCD04077665
CAS Number: 662164-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty