Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(4-((5-Chloro-4-((4-chlorophenyl)amino)pyrimidin-2-yl)amino)-1H-pyrazol-1-yl)-N-hydroxyhexanamide 98%

Supplier: AMBEED, INC
Warning

A1155022-10MG A1155022-1MG A1155022-25MG A1155022-5MG
76674-852EA 52 USD InStock
76674-852 76674-854 76674-856 76674-858
6-(4-((5-Chloro-4-((4-chlorophenyl)amino)pyrimidin-2-yl)amino)-1H-pyrazol-1-yl)-N-hydroxyhexanamide 98%
6-(4-((5-Chloro-4-((4-chlorophenyl)amino)pyrimidin-2-yl)amino)-1H-pyrazol-1-yl)-N-hydroxyhexanamide
Precaución: for Research Use Only


Formula: C₁₉H₂₁Cl₂N₇O₂
MW: 450.32 g/mol
CAS Number: 2284621-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty