Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,7-Diethylnonane-4,6-dione 97%

Supplier: AMBEED, INC
Warning

A1156055-100MG A1156055-1G A1156055-250MG A1156055-25G A1156055-5G
76674-960EA 6 USD InStock
76674-960 76674-962 76674-964 76674-966 76674-968
3,7-Diethylnonane-4,6-dione 97%
3,7-Diethylnonane-4,6-dione
Precaución: for Research Use Only


Formula: C₁₃H₂₄O₂
MDL Number: MFCD30738768
CAS Number: 872802-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty