Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylisonicotinic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1156584-100MG A1156584-1G A1156584-250MG
76675-064EA 89 USD InStock
76675-064 76675-066 76675-068
2,3-Dimethylisonicotinic acid hydrochloride 97%
2,3-Dimethylisonicotinic acid hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀ClNO₂
CAS Number: 2442597-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty