Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dipropoxyterephthalohydrazide 98%

Supplier: AMBEED, INC
Warning

A1156829-100MG A1156829-1G A1156829-250MG A1156829-5G
76675-134EA 20 USD InStock
76675-134 76675-136 76675-138 76675-140
2,5-Dipropoxyterephthalohydrazide 98%
2,5-Dipropoxyterephthalohydrazide
Precaución: for Research Use Only


Formula: C₁₄H₂₂N₄O₄
CAS Number: 2143456-42-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty