Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5'-(4-Carboxyphenyl)-2'-methyl-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A1156830-100MG A1156830-1G A1156830-250MG
76675-142EA 16 USD InStock
76675-142 76675-144 76675-146
5'-(4-Carboxyphenyl)-2'-methyl-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid 98%
5'-(4-Carboxyphenyl)-2'-methyl-[1,1':3',1''-terphenyl]-4,4''-dicarboxylic acid
Precaución: for Research Use Only


Formula: C₂₈H₂₀O₆
MW: 452.4548 g/mol
MDL Number: MFCD34165570
CAS Number: 1774401-31-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty