Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic acid, 2',5'-dimethyl- 98%

Supplier: AMBEED, INC
Warning

A1156856-100MG A1156856-1G A1156856-250MG A1156856-5G
76675-182EA 15 USD InStock
76675-182 76675-184 76675-186 76675-188
[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic acid, 2',5'-dimethyl- 98%
[1,1':4',1''-Terphenyl]-4,4''-dicarboxylic acid, 2',5'-dimethyl-
Precaución: for Research Use Only


Formula: C₂₂H₁₈O₄
MDL Number: MFCD32666997
CAS Number: 115213-33-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty