Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Di(pyridin-4-yl)naphthalene 98%

Supplier: AMBEED, INC
Warning

A1156880-100MG A1156880-1G A1156880-250MG A1156880-5G
76675-232EA 44 USD InStock
76675-232 76675-234 76675-236 76675-238
1,4-Di(pyridin-4-yl)naphthalene 98%
1,4-Di(pyridin-4-yl)naphthalene
Precaución: for Research Use Only


Formula: C₂₀H₁₄N₂
CAS Number: 1429342-63-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty