Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N1,N3,N5-Tri(pyridin-4-yl)benzene-1,3,5-tricarboxamide 98%

Supplier: AMBEED, INC
Warning

A1156885-100MG A1156885-1G A1156885-250MG A1156885-5G
76675-248EA 22 USD InStock
76675-248 76675-250 76675-252 76675-254
N1,N3,N5-Tri(pyridin-4-yl)benzene-1,3,5-tricarboxamide 98%
N1,N3,N5-Tri(pyridin-4-yl)benzene-1,3,5-tricarboxamide
Precaución: for Research Use Only


Formula: C₂₄H₁₈N₆O₃
MDL Number: MFCD32068205
CAS Number: 725274-03-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty