Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1-isocyanato-3-(trifluoromethyl)benzene 98%

Supplier: AMBEED, INC
Danger

A1157815-1G A1157815-250MG A1157815-5G
76675-336EA 619 USD InStock
76675-336 76675-338 76675-340
2-Chloro-1-isocyanato-3-(trifluoromethyl)benzene 98%
2-Chloro-1-isocyanato-3-(trifluoromethyl)benzene
Precaución: for Research Use Only


Formula: C₈H₃ClF₃NO
MDL Number: MFCD28359385
CAS Number: 88330-63-6
UN: 2206
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty