Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2R,5R)-5-Benzyl-3-methyl-2-(5-methylfuran-2-yl)imidazolidin-4-one 98%

Supplier: AMBEED, INC
Danger

A1158110-100MG A1158110-250MG
76675-352EA 58 USD InStock
76675-352 76675-354
(2R,5R)-5-Benzyl-3-methyl-2-(5-methylfuran-2-yl)imidazolidin-4-one 98%
(2R,5R)-5-Benzyl-3-methyl-2-(5-methylfuran-2-yl)imidazolidin-4-one
Precaución: for Research Use Only


Formula: C₁₆H₁₈N₂O₂
MDL Number: MFCD08457654
CAS Number: 877303-84-9
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty