Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzoic acid, 4,4'-(1-hydroxy-2-oxo-1,2-ethanediyl)bis-, 1,1'-methyl ester 98%

Supplier: AMBEED, INC
Warning

A1158605-100MG A1158605-1G A1158605-250MG
76675-374EA 21 USD InStock
76675-374 76675-376 76675-378
Benzoic acid, 4,4'-(1-hydroxy-2-oxo-1,2-ethanediyl)bis-, 1,1'-methyl ester 98%
Benzoic acid, 4,4'-(1-hydroxy-2-oxo-1,2-ethanediyl)bis-, 1,1'-methyl ester
Precaución: for Research Use Only


Formula: C₁₈H₁₆O₆
MDL Number: MFCD31758394
CAS Number: 40742-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty