Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

o-Xylylenediphosphonic acid 98%

Supplier: AMBEED, INC
Danger

A1158663-1G A1158663-250MG A1158663-5G
76675-380EA 17 USD InStock
76675-380 76675-382 76675-384
o-Xylylenediphosphonic acid 98%
o-Xylylenediphosphonic acid
Precaución: for Research Use Only


Formula: C₈H₁₂O₆P₂
MDL Number: MFCD22200404
CAS Number: 42104-58-5
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty