Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate 97%

Supplier: AMBEED, INC
Warning

A1158917-1G A1158917-25G A1158917-5G
76675-452EA 20 USD InStock
76675-452 76675-454 76675-456
2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate 97%
2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₄
MDL Number: MFCD31715034
CAS Number: 254900-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty