Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(acetonitrile)pentamethylcyclopentadienylcobalt(II) hexafluoroantimonate 97%

Supplier: AMBEED, INC
Danger

A1159429-100MG A1159429-50MG A1159429-250MG
76675-514EA 69 USD InStock
76675-514 76675-516 77620-296
Tris(acetonitrile)pentamethylcyclopentadienylcobalt(II) hexafluoroantimonate 97%
Tris(acetonitrile)pentamethylcyclopentadienylcobalt(II) hexafluoroantimonate
Precaución: for Research Use Only


Formula: C₁₆H₂₄CoF₆N₃Sb
CAS Number: 1642132-26-0
UN: 3467
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty