Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1159447-10G A1159447-1G A1159447-250MG A1159447-5G
76675-518EA 105 USD InStock
76675-518 76675-520 76675-522 76675-524
(2-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride 97%
(2-((Dimethylamino)methyl)phenyl)boronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₅BClNO₂
MDL Number: MFCD27952814
CAS Number: 2828431-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty