Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-5-(piperidin-4-yl)-1,3,4-oxadiazole hydrobromide 97%

Supplier: AMBEED, INC
Warning

A1159560-1G A1159560-250MG
76675-540EA 261 USD InStock
76675-540 76675-542
2-Methyl-5-(piperidin-4-yl)-1,3,4-oxadiazole hydrobromide 97%
2-Methyl-5-(piperidin-4-yl)-1,3,4-oxadiazole hydrobromide
Precaución: for Research Use Only


Formula: C₈H₁₄BrN₃O
CAS Number: 2442597-61-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty