Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Azabicyclo[3.3.1]nonane n-oxyl 98%

Supplier: AMBEED, INC
Danger

A1162957-100MG A1162957-1G A1162957-250MG A1162957-5G
76675-592EA 42 USD InStock
76675-592 76675-594 76675-596 76675-598
9-Azabicyclo[3.3.1]nonane n-oxyl 98%
9-Azabicyclo[3.3.1]nonane n-oxyl
Precaución: for Research Use Only


Formula: C₈H₁₄NO
MW: 140.2029 g/mol
MDL Number: MFCD23704408
CAS Number: 31785-68-9
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty