Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(1,2-Diphenylethene-1,2-diyl)diphenol 97%

Supplier: AMBEED, INC
Warning

A1164714-1G A1164714-250MG A1164714-25G A1164714-5G
76675-682EA 47 USD InStock
76675-682 76675-684 76675-686 76675-688
4,4'-(1,2-Diphenylethene-1,2-diyl)diphenol 97%
4,4'-(1,2-Diphenylethene-1,2-diyl)diphenol
Precaución: for Research Use Only


Formula: C₂₆H₂₀O₂
MDL Number: MFCD28976010
CAS Number: 68578-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty