Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-([1,1'-Biphenyl]-4,4'-diyl)diacetonitrile 98%

Supplier: AMBEED, INC
Danger

A1164720-100MG A1164720-1G
76675-690EA 15 USD InStock
76675-690 76675-692
2,2'-([1,1'-Biphenyl]-4,4'-diyl)diacetonitrile 98%
2,2'-([1,1'-Biphenyl]-4,4'-diyl)diacetonitrile
Precaución: for Research Use Only


Formula: C₁₆H₁₂N₂
MDL Number: MFCD00051458
CAS Number: 7255-83-6
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty