Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclobutyl-3-(2,6-dimethylphenyl)-7-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one 99%

Supplier: AMBEED, INC
Warning

A1164822-10MG A1164822-1MG A1164822-50MG A1164822-5MG
76675-704EA 59 USD InStock
76675-704 76675-706 76675-708 76675-710
1-Cyclobutyl-3-(2,6-dimethylphenyl)-7-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one 99%
1-Cyclobutyl-3-(2,6-dimethylphenyl)-7-((2-methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one
Precaución: for Research Use Only


Formula: C₃₀H₃₇N₇O₂
MDL Number: MFCD32173438
CAS Number: 2172617-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty