Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(3-(Difluoromethyl)-2-fluorophenyl)ethanamine 97%

Supplier: AMBEED, INC
Danger

A1164913-1G A1164913-5G A1164913-100MG
76675-726EA 97 USD InStock
76675-726 76675-728 77620-348
(R)-1-(3-(Difluoromethyl)-2-fluorophenyl)ethanamine 97%
(R)-1-(3-(Difluoromethyl)-2-fluorophenyl)ethanamine
Precaución: for Research Use Only


Formula: C₉H₁₀F₃N
MDL Number: MFCD22504810
CAS Number: 1389852-29-2
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty