Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

m-Xylene-4-sulfonic acid hydrate 98% (by HPLC) (contains ≤10% H₂O)

Supplier: AMBEED, INC
Warning

A1165015-100G A1165015-500G
76675-738EA 11 USD InStock
76675-738 76675-740
m-Xylene-4-sulfonic acid hydrate 98% (by HPLC) (contains ≤10% H₂O)
m-Xylene-4-sulfonic acid hydrate
Precaución: for Research Use Only


Formula: C₈H₁₀O₃S
MDL Number: MFCD00007483
CAS Number: 88-61-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty