Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polypropylene glycol ∼ Mn 4000

Supplier: AMBEED, INC
Warning

A1165276-1000G A1165276-100G A1165276-500G
76675-778EA 17 USD InStock
76675-778 76675-780 76675-782
Polypropylene glycol ∼ Mn 4000
Polypropylene glycol
Precaución: for Research Use Only


Formula: H(OCH₂CH₂CH₂)nOH
MDL Number: MFCD00084446
CAS Number: 25322-69-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty