Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-Methyl 2-(2-(((1,3-dioxoisoindolin-2-yl)oxy)methyl)phenyl)-2-(methoxyimino)acetate 97%

Supplier: AMBEED, INC
Danger

A1165830-100MG A1165830-1G A1165830-250MG A1165830-5G
76675-836EA 59 USD InStock
76675-836 76675-838 76675-840 76675-842
(E)-Methyl 2-(2-(((1,3-dioxoisoindolin-2-yl)oxy)methyl)phenyl)-2-(methoxyimino)acetate 97%
(E)-Methyl 2-(2-(((1,3-dioxoisoindolin-2-yl)oxy)methyl)phenyl)-2-(methoxyimino)acetate
Precaución: for Research Use Only


Formula: C₁₉H₁₆N₂O₆
CAS Number: 156581-14-5
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty