Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-((S)-1-(((2S,3S)-1-((6-Amino-6-oxohexyl)amino)-3-methyl-1-oxopentan-2-yl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)hexanamide acetate 98%

Supplier: AMBEED, INC
Warning

A1165988-10MG A1165988-1MG A1165988-25MG
76675-864EA 60 USD InStock
76675-864 76675-866 76675-868
N-((S)-1-(((2S,3S)-1-((6-Amino-6-oxohexyl)amino)-3-methyl-1-oxopentan-2-yl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)hexanamide acetate 98%
N-((S)-1-(((2S,3S)-1-((6-Amino-6-oxohexyl)amino)-3-methyl-1-oxopentan-2-yl)amino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)hexanamide acetate
Precaución: for Research Use Only


Formula: C₂₉H₄₈N₄O₇
CAS Number: 2453297-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty