Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-(Hydroxymethyl)phenyl)-2-methylpropanenitrile 97%

Supplier: AMBEED, INC
Warning

A1166499-1G A1166499-250MG
76675-914EA 277 USD InStock
76675-914 76675-916
2-(4-(Hydroxymethyl)phenyl)-2-methylpropanenitrile 97%
2-(4-(Hydroxymethyl)phenyl)-2-methylpropanenitrile
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO
MDL Number: MFCD19695275
CAS Number: 91132-15-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty