Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(tert-Butyl)-2-ethoxybenzenesulfonamide 97%

Supplier: AMBEED, INC
Warning

A1168305-100MG A1168305-1G A1168305-250MG
76676-064EA 17 USD InStock
76676-064 76676-066 76676-068
5-(tert-Butyl)-2-ethoxybenzenesulfonamide 97%
5-(tert-Butyl)-2-ethoxybenzenesulfonamide
Precaución: for Research Use Only


Formula: C₁₂H₁₉NO₃S
MW: 257.3492 g/mol
MDL Number: MFCD12402660
CAS Number: 1206134-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty