Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Di(adamantan-1-yl)(2-((triisopropylsilyl)oxy)phenyl)phosphine 97%

Supplier: AMBEED, INC
Warning

A1168687-100MG A1168687-1G A1168687-250MG
76676-132EA 105 USD InStock
76676-132 76676-134 76676-136
Di(adamantan-1-yl)(2-((triisopropylsilyl)oxy)phenyl)phosphine 97%
Di(adamantan-1-yl)(2-((triisopropylsilyl)oxy)phenyl)phosphine
Precaución: for Research Use Only


Formula: C₃₅H₅₅OPSi
MW: 550.8699 g/mol
MDL Number: MFCD23098736
CAS Number: 1384966-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty