Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Bromothiophen-2-yl)ethanamine hydrochloride 97%

Supplier: AMBEED, INC
Danger

A1174931-100MG A1174931-1G A1174931-250MG A1174931-5G
76676-190EA 11 USD InStock
76676-190 76676-192 76676-194 76676-196
1-(4-Bromothiophen-2-yl)ethanamine hydrochloride 97%
1-(4-Bromothiophen-2-yl)ethanamine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₉BrClNS
MW: 242.5644 g/mol
MDL Number: MFCD32197986
CAS Number: 2244906-14-5
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty