Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-((2R,3R)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)pyridine 97%

Supplier: AMBEED, INC
Warning

A1175207-100MG A1175207-250MG
76676-242EA 120 USD InStock
76676-242 77620-440
2-((2R,3R)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)pyridine 97%
2-((2R,3R)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)pyridine
Precaución: for Research Use Only


Formula: C₃₀H₂₆NOP
MDL Number: MFCD32641187
CAS Number: 1542796-14-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty