Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-((S)-(2-(diphenylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide phenyl)(phenyl)methyl)-2- methylpropane-2-sulfinamide 97%

Supplier: AMBEED, INC
Warning

A1175252-250MG A1175252-50MG A1175252-100MG
76676-270EA 240 USD InStock
76676-270 76676-272 77620-448
(R)-N-((S)-(2-(diphenylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide phenyl)(phenyl)methyl)-2- methylpropane-2-sulfinamide 97%
(R)-N-((S)-(2-(diphenylphosphanyl)phenyl)(phenyl)methyl)-2-methylpropane-2-sulfinamide phenyl)(phenyl)methyl)-2- methylpropane-2-sulfinamide


Formula: C₂₉H₃₀NOPS
MW: 471.5934 g/mol
MDL Number: MFCD32697141
CAS Number: 1595319-97-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty