Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine 98%

Supplier: AMBEED, INC
Warning

A1175445-100MG A1175445-1G A1175445-250MG A1175445-5G
76676-300EA 16 USD InStock
76676-300 76676-302 76676-304 76676-306
4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine 98%
4-Chloro-6-(4-chlorophenyl)-2-phenylpyrimidine
Precaución: for Research Use Only


Formula: C₁₆H₁₀Cl₂N₂
MDL Number: MFCD30536778
CAS Number: 1354749-13-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty