Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(dibenzylideneacetone)diplatinum(0) 98%

Supplier: AMBEED, INC
Warning

A1175867-100MG A1175867-1G A1175867-250MG
76676-334EA 111 USD InStock
76676-334 76676-336 76676-338
Tris(dibenzylideneacetone)diplatinum(0) 98%
Tris(dibenzylideneacetone)diplatinum(0)
Precaución: for Research Use Only


Formula: C₅₁H₄₂O₃Pt₂
MW: 1093.0334 g/mol
MDL Number: MFCD03426034
CAS Number: 63782-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty