Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,2'-bipyridyl) bis [2- (4-tert-butylphenyl) pyridine] iridium (III) hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A1175916-100MG A1175916-1G A1175916-50MG A1175916-250MG
76676-348EA 68 USD InStock
76676-348 76676-350 76676-352 77620-482
(2,2'-bipyridyl) bis [2- (4-tert-butylphenyl) pyridine] iridium (III) hexafluorophosphate 98%
(2,2'-bipyridyl) bis [2- (4-tert-butylphenyl) pyridine] iridium (III) hexafluorophosphate
Precaución: for Research Use Only


Formula: C₄₀H₄₀F₆IrN₄P
CAS Number: 1352428-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty